Merck
  • A novel anticancer pharmacological agent targeting mitochondrial complex I.
A novel anticancer pharmacological agent targeting mitochondrial complex I.
Trends in pharmacological sciences (2022-04-07)
Gabriela Reyes-Castellanos, Alice Carrier
ABSTRACT
Targeting metabolic reprogramming has proven successful in oncology, but this field requires better identification of drugs that inhibit mitochondrial metabolism in cancer cells. Recent work from Dr Wolf's group reveals that the primary target of the antitumor compound SMIP004-7 is mitochondrial complex I (NDUFS2 subunit), inhibition of which promotes anticancer immune surveillance.
Sigma-Aldrich
SMIP004-7, ≥98% (HPLC)