Merck
  • Review on application of perylene diimide (PDI)-based materials in environment: Pollutant detection and degradation.
Review on application of perylene diimide (PDI)-based materials in environment: Pollutant detection and degradation.
The Science of the total environment (2021-03-28)
Wenwu Zhou, Guo Liu, Bing Yang, Qiuyi Ji, Weiming Xiang, Huan He, Zhe Xu, Chengdu Qi, Shiyin Li, Shaogui Yang, Chenmin Xu
ABSTRACT
Environment pollution is getting serious and various poisonous contaminants with chemical durability, biotoxicity and bioaccumulation have been widespreadly discovered in municipal wastewaters and surface water. The detection and removal of pollutants show great significance for the protection of human health and other organisms. Due to its distinctive physical and chemical properties, perylene diimide (PDI) has received widespread attention from different research fields, especially in the area of environment. In this review, a comprehensive summary of the development of PDI-based materials in fluorescence detection and advanced oxidation technology for environment was introduced. Firstly, we chiefly presented the recent progress about the synthesis of PDI and PDI-based nanomaterials. Then, their application in fluorescence detection for environment was presented and categorized, principally including the detection of heavy metal ions, harmful anions and organic contaminants in the environment. In addition, the application of PDI and PDI-based materials in different advanced oxidation technologies for environment, such as photocatalysis, photoelectrocatalysis, Fenton and Fenton-like reaction and persulfate activation, was also summarized. At last, the challenges and future prospects of PDI-based materials in environmental applications were discussed. This review focuses on presenting the practical applications of PDI and PDI-based materials as fluorescent probes or catalysts (especially photocatalysts) in the detection of hazardous substances or catalytic elimination of organic contaminants. The contents are aimed at supplying the researchers with a deeper understanding of PDI and PDI-based materials and encouraging their further development in environmental applications.
Sigma-Aldrich
3,4,9,10-Perylenetetracarboxylic dianhydride
Pricing and availability is not currently available.