Merck
  • Cab45 deficiency leads to the mistargeting of progranulin and prosaposin and aberrant lysosomal positioning.
Cab45 deficiency leads to the mistargeting of progranulin and prosaposin and aberrant lysosomal positioning.
Traffic (Copenhagen, Denmark) (2022-11-19)
Mai Ly Tran, Johanna Tüshaus, Yeongho Kim, Bulat R Ramazanov, Swathi Devireddy, Stefan F Lichtenthaler, Shawn M Ferguson, Julia von Blume
ABSTRACT
The trans-Golgi Network (TGN) sorts molecular "addresses" and sends newly synthesized proteins to their destination via vesicular transport carriers. Despite the functional significance of packaging processes at the TGN, the sorting of soluble proteins remains poorly understood. Recent research has shown that the Golgi resident protein Cab45 is a significant regulator of secretory cargo sorting at the TGN. Cab45 oligomerizes upon transient Ca2+ influx, recruits soluble cargo molecules (clients), and packs them in sphingomyelin-rich transport carriers. However, the identity of client molecules packed into Cab45 vesicles is scarce. Therefore, we used a precise and highly efficient secretome analysis technology called hiSPECs. Intriguingly, we observed that Cab45 deficient cells manifest hypersecretion of lysosomal hydrolases. Specifically, Cab45 deficient cells secrete the unprocessed precursors of prosaposin (PSAP) and progranulin (PGRN). In addition, lysosomes in these cells show an aberrant perinuclear accumulation suggesting a new role of Cab45 in lysosomal positioning. This work uncovers a yet unknown function of Cab45 in regulating lysosomal function.
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), agarose conjugate, Type VI, saline suspension
Pricing and availability is not currently available.