Merck
  • A rapid, economical, and reproducible method for human serum delipidation and albumin and IgG removal for proteomic analysis.
A rapid, economical, and reproducible method for human serum delipidation and albumin and IgG removal for proteomic analysis.
Methods in molecular biology (Clifton, N.J.) (2006-12-19)
Qin Fu, Diane E Bovenkamp, Jennifer E Van Eyk
ABSTRACT
Serum is a readily available source for diagnostic assays, but the identification of disease-specific serum biomarkers has been impeded by the dominance of human serum albumin (HSA) and immunoglobulin G (IgG) in the serum proteome. Therefore, in order to observe lower-abundance serum proteins, removal or depletion of at least these two proteins is required. However, the depletion method needs to be inexpensive and reproducible. We describe such a protocol that combines delipidation by centrifugation, IgG removal with Protein G Sepharose, and HSA depletion with sodium chloride/ethanol precipitation. The protocol is streamlined to increase reproducibility and is compatible with many proteomic platforms, including two-dimensional gel electrophoresis, and high-performance liquid chromatography either offline or coupled online with a mass spectrometer. The reproducible depletion of lipids, IgG, and HSA permits a higher load of the remaining serum proteins, facilitating the identification of disease biomarkers.
Sigma-Aldrich
Sera human, frozen liquid
Pricing and availability is not currently available.