Merck
  • Effects of beta-glucanase and xylanase supplementation on gastrointestinal digestive enzyme activities of weaned piglets fed a barley-based diet.
Effects of beta-glucanase and xylanase supplementation on gastrointestinal digestive enzyme activities of weaned piglets fed a barley-based diet.
Journal of animal physiology and animal nutrition (2008-05-22)
C L Fan, X Y Han, Z R Xu, L J Wang, L R Shi
ABSTRACT
The effects of supplementing a barley-based diet for weaned piglets withexogenous beta-glucanase and xylanase on gastrointestinal digestiveenzyme activities were investigated. Thirty-six cross-bred weaned pigletswere randomly assigned to two groups with three pens based on sexand mass. Each group was fed on the diet based on barley with or withoutadded beta-glucanase and xylanase (0.15%) for a 4-week period. Theresults showed that enzyme supplementation improved growth performanceof piglets significantly (p < 0.05), but had no effect (p = 0.091)on average daily feed intake. The results also showed that supplementationof beta-glucanase and xylanase had no effect on pepsin activity in gastriccontents but slightly decreased (p = 0.092) the pepsin activity ingastric mucosa. Meanwhile, no effect of enzyme supplementation ontrypsin activity in duodenal contents was observed. However, the activitiesof amylase and lipase in duodenal contents were significantly(p < 0.05) decreased, whereas the activities of maltase, sucrase andgamma-glutamyl transpeptidase (gamma-GT) in jejunal and ileal mucosa wereenhanced significantly (p < 0.05). The improvement of disaccharidaseand gamma-GT activity may be attributed to the positive impacts of exogenousenzymes on digestion and absorption of the nutrients. In conclusion,the current results indicated that supplementation with enzymes in barley-based diets could improve the growth performance of piglets,decrease the activities of amylase and lipase in duodenal contents andincrease the activities of disaccharidase and gamma-GT in jejunal and ilealmucosa.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)