Merck
  • Immunolocalization of dentin matrix protein-1 in human primary teeth treated with different pulp capping materials.
Immunolocalization of dentin matrix protein-1 in human primary teeth treated with different pulp capping materials.
Journal of biomedical materials research. Part B, Applied biomaterials (2015-02-14)
Natalino Lourenço Neto, Nádia C T Marques, Ana Paula Fernandes, Camila O Rodini, Vivien T Sakai, Ruy Cesar C Abdo, Maria Aparecida A M Machado, Carlos F Santos, Thais M Oliveira
ABSTRACT
The aim of this study was to evaluate the immunolocalization of dentin matrix protein (DMP)-1 in human primary teeth treated with different pulp capping materials. Twenty-five primary molars were divided into the following groups: formocresol (FC), calcium hydroxide (CH), mineral trioxide aggregate (MTA), corticosteroid/antibiotic solution + CH (O + CH), and Portland cement (PC), and all received conventional pulpotomy treatment. The teeth at the regular exfoliation period were extracted for histological analysis and immunolocalization of DMP-1. Statistical analysis was performed using the χ(2) test (p < 0.05). Histological analysis revealed statistically significant differences in the comparison among the groups through the use of a score system regarding the presence of hard tissue barrier, odontoblastic layer, and internal resorption, but not regarding pulp calcification. Immunohistochemical analysis showed immunostaining for DMP-1 in groups CH, MTA, O + CH, and PC. Internal resorption was observed in the groups FC and CH. MTA and PC showed pulp repair without inflammation and with the presence of hard tissue barrier. DMP-1 immunostaining was higher for MTA and PC, confirming the reparative and bioinductive capacity of these materials.
Sigma-Aldrich
Anti-DMP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.