- Transcriptome modulation following administration of luteolin to bleomycin-etoposide-cisplatin chemotherapy on rat LC540 tumor Leydig cells.
Transcriptome modulation following administration of luteolin to bleomycin-etoposide-cisplatin chemotherapy on rat LC540 tumor Leydig cells.
Leydig cell tumours represent 1%-3% of all cases of testicular tumours in men. Such tumours respond poorly to radiation or chemotherapy, including bleomycin-etoposide-cisplatin (BEP) combinatorial therapy. In this study, we investigated an alternative approach involving luteolin to improve the efficacy of chemotherapy. LC540 tumour Leydig cells were treated with BEP (bleomycin 40 µg/ml, etoposide 4 µg/ml, cisplatin 8 µg/ml) and/or luteolin 10 µM for comparison with DMSO-treated cells. We performed a transcriptome analysis using RNA-Seq to characterise changes in biological processes and signalling pathways. Treatments of LC540 tumour Leydig cells with luteolin significantly decreased the expression of genes involved in cholesterol biosynthesis, while increasing the expression of genes related to glutathione conjugation (p < .05). Genes being significantly upregulated in response to BEP treatment were involved in the response to toxic substances and transcriptional regulation. Oppositely, genes being significantly downregulated by BEP treatment were enriched for intracellular signal transduction, cell migration, cell adhesion, reproductive system development and cholesterol biosynthesis. BEP chemotherapy proved to be effective in increasing gene expression related to apoptosis of tumour Leydig cells. However, addition of luteolin to BEP treatment had no other effects on biological processes or pathways related to cancer treatment.