Merck
Long-Term Culture of Nephron Progenitor Cells Ex Vivo.
Methods in molecular biology (Clifton, N.J.) (2019-02-12)
Aaron C Brown, Ashwani K Gupta, Leif Oxburgh
ABSTRACT
Nephrons differentiate from the cap mesenchyme of the fetal kidney. Nephron progenitor cells that populate the cap mesenchyme efficiently balance self-renewal and epithelial differentiation to enable repeated rounds of nephron formation during development. Here we describe a method to isolate and propagate these cells from the embryonic mouse kidney. Using this method, nephron progenitor cells from a single litter of mice can be propagated to hundreds of millions of cells that express appropriate markers of the undifferentiated state and retain epithelial differentiation capacity in vitro.
Sigma-Aldrich
Y-27632, InSolution, ≥95%, reversible, inhibitor of Rho kinases
Pricing and availability is not currently available.