Merck
  • Optimization of Capture ELISAs for Chicken Cytokines Using Commercially Available Antibodies.
Optimization of Capture ELISAs for Chicken Cytokines Using Commercially Available Antibodies.
Animals : an open access journal from MDPI (2022-11-12)
Paulina Krzysica, Loes Verhoog, Sonja de Vries, Coen Smits, Huub F J Savelkoul, Edwin Tijhaar
ABSTRACT
Cytokines like interferon (IFN)-γ, interleukin (IL)-2, IL-6, IL-10, and IL-12p40 are important biomarkers for characterizing the nature and strength of immune responses. It is important to be able to quantify the cytokines at the protein level in biological samples. Quantification of chicken cytokines is generally performed on the level of messenger RNA (mRNA) by quantitative polymerase chain reaction (qPCR) because very few capture ELISAs for the quantification of chicken cytokine proteins are commercially available. Here, we describe the optimization and validation of capture ELISAs for chicken IL-2, IL-6, IL-10, IL-12p40, and IFN-γ using commercially available antibodies and reagents. First, we determined the optimal concentrations of the antibodies. We then verified the ELISAs' performance and established that the lower limit of detection (LLOD) for all cytokines was below 32 pg/mL. The ELISAs show the same binding characteristics for recombinant and native cytokines (parallelism was <15.2% CV). Values for inter-assay variation were consistently low and mostly <20% CV. Overall, the optimized capture ELISAs are sensitive (<32 pg/mL) and reliable tools to quantify chicken cytokines. These ELISAs can easily and inexpensively be utilized in any immunological lab and may therefore have wide applicability in immunological research for poultry.
Product Description
Roche
Bovine Serum Albumin, ≥98.5%, New Zealand origin
Pricing and availability is not currently available.
Sigma-Aldrich
Lectin from Phytolacca americana (pokeweed), lyophilized powder
Pricing and availability is not currently available.