Merck
  • Vibrio cholerae O1 infection induces proinflammatory CD4+ T-cell responses in blood and intestinal mucosa of infected humans.
Vibrio cholerae O1 infection induces proinflammatory CD4+ T-cell responses in blood and intestinal mucosa of infected humans.
Clinical and vaccine immunology : CVI (2011-06-24)
Alison Kuchta, Taibur Rahman, Erica L Sennott, Taufiqur R Bhuyian, Taher Uddin, Rasheduzzaman Rashu, Fahima Chowdhury, Ashraf I Kahn, Mohammad Arifuzzaman, Ana A Weil, Michael Podolsky, Regina C LaRocque, Edward T Ryan, Stephen B Calderwood, Firdausi Qadri, Jason B Harris
ABSTRACT
Vibrio cholerae O1 is a noninvasive enteric pathogen and serves as a model for studies of mucosal immunity. Although symptomatic V. cholerae infection induces durable protection against subsequent disease, vaccination with oral killed whole-cell V. cholerae stimulates less long-lasting protection against cholera. In this study, we demonstrated that cholera induces an early proinflammatory cellular immune response that results in priming of Th1- and Th17-type cytokine responses to ex vivo antigenic stimulation and an increase in the ratio of Th1 to Th2 CD4(+) T-cell responses. Comparable priming of Th1 and Th17 responses, with an increased ratio of Th1 to Th2 CD4(+) T-cell responses, was not observed in subjects who received two doses of the oral cholera vaccine Dukoral (a whole-cell cholera toxin B subunit containing [WC-CTB] vaccine). These findings suggest that natural V. cholerae infection induces an early, proinflammatory cellular immune response, despite the apparent lack of clinical signs of inflammation. The failure of the WC-CTB vaccine to activate equivalent, CD4(+) T-cell responses is a potential explanation for the shorter duration of protection following immunization with this vaccine. Additional studies are needed to determine whether these early T-cell-mediated events predict the subsequent duration of immunologic memory.
Sigma-Aldrich
Hanks′ Balanced Salt Solution 10x, Without calcium chloride, magnesium sulfate and sodium bicarbonate, 10 ×, liquid, sterile-filtered, suitable for cell culture
Pricing and availability is not currently available.