Merck
  • Lewis Acid Catalyzed Cyclization Reactions of Ethenetricarboxylates via Intramolecular Hydride Transfer.
Lewis Acid Catalyzed Cyclization Reactions of Ethenetricarboxylates via Intramolecular Hydride Transfer.
The Journal of organic chemistry (2017-06-13)
Shoko Yamazaki, Taku Naito, Mamiko Niina, Kiyomi Kakiuchi
ABSTRACT
Catalytic cyclization of amides of ethenetricarboxylate bearing ether and acetal groups has been examined. The reaction of the amides bearing cyclic ether and acetal groups in the presence of Lewis acid such as Sc(OTf)3 gave spirocyclic piperidine derivatives as major products. The cyclized products may be formed via intramolecular hydride transfer. The reaction mechanism was examined by the DFT calculations. The scope and limitations of the hydride transfer/cyclization reactions of amides of ethenetricarboxylates was investigated, and morpholine formation by intramolecular oxy-Michael addition was also found.
Sigma-Aldrich
(±)-α-Methoxyphenylacetic acid, crystalline