Merck
  • Selective quantification of the 22-kDa isoform of human growth hormone 1 in serum and plasma by immunocapture and LC-MS/MS.
Selective quantification of the 22-kDa isoform of human growth hormone 1 in serum and plasma by immunocapture and LC-MS/MS.
Analytical and bioanalytical chemistry (2022-07-16)
Bas Sleumer, Martijn van Faassen, Michel J Vos, Rainer Bischoff, Ido P Kema, Nico C van de Merbel
ABSTRACT
The human growth hormone GH1 (22 kDa) is a commonly measured biomarker for diagnosis and during treatment of growth disorders, but its quantification by ligand binding assays may be compromised by the occurrence of a number of isoforms. These can interfere in the assays and lead to differences in results between laboratories and potentially even in the treatment of patients. We present an LC-MS/MS method that is able to distinguish the major growth hormone isoform (GH1, 22 kDa) from other isoforms and quantify it without any interference across the clinically relevant concentration range of 0.5 to 50 ng/mL. Analysis involves purification of a 100-µL serum sample by immunocapture using an anti-GH-directed antibody, tryptic digestion, and LC-MS/MS quantification of an isoform-specific signature peptide for GH1 (22 kDa). A tryptic peptide occurring in all GH isoforms is monitored in the same 16-min analytical run as a read-out for total GH. Stable-isotope-labeled forms of these two peptides are included as internal standards. Full validation of the method according to recent guidelines, against a recombinant form of the analyte in rat plasma calibrators, demonstrated intra-assay and inter-assay imprecision below 6% across the calibration range for both signature peptides and recoveries between 94 and 102%. An excellent correlation was found between nominal and measured concentrations of the WHO reference standard for GH1 (22 kDa). Addition of up to 1000 ng/mL biotin or the presence of a 100-fold excess of GH binding protein did not affect the measurement. Equivalent method performance was found for analysis of GH in serum, EDTA, and heparin plasma. Analyte stability was demonstrated during all normal sample storage conditions. Comparison with the IDS-iSYS GH immunoassay showed a good correlation with the LC-MS/MS method for the isoform-specific signature peptide, but a significant positive bias was observed for the LC-MS/MS results of the peptide representing total GH. This seems to confirm the actual occurrence of other GH isoforms in serum. Finally, in serum from pregnant individuals, no quantifiable GH1 (22 kDa) was found, but relatively high concentrations of total GH.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Pricing and availability is not currently available.
Roche
Biotin Protein Labeling Kit, sufficient for 5 labeling reactions, kit of 1 (5 components), suitable for immunoblotting, suitable for ELISA
Pricing and availability is not currently available.
Sigma-Aldrich
Trypsin from porcine pancreas, Type IX-S, lyophilized powder, 13,000-20,000 BAEE units/mg protein
Pricing and availability is not currently available.