Merck
  • Chemoradiation induces epithelial-to-mesenchymal transition in esophageal adenocarcinoma.
Chemoradiation induces epithelial-to-mesenchymal transition in esophageal adenocarcinoma.
International journal of cancer (2019-04-25)
Anne Steins, Eva A Ebbing, Aafke Creemers, Amber P van der Zalm, Rajni A Jibodh, Cynthia Waasdorp, Sybren L Meijer, Otto M van Delden, Kausilia K Krishnadath, Maarten C C M Hulshof, Roelof J Bennink, Cornelis J A Punt, Jan Paul Medema, Maarten F Bijlsma, Hanneke W M van Laarhoven
ABSTRACT
Multimodality treatment has advanced the outcome of esophageal adenocarcinoma (EAC), but overall survival remains poor. Therapeutic pressure activates effective resistance mechanisms and we characterized these mechanisms in response to the currently used neoadjuvant treatment against EAC: carboplatin, paclitaxel and radiotherapy. We developed an in vitro approximation of this regimen and applied it to primary patient-derived cultures. We observed a heterogeneous epithelial-to-mesenchymal (EMT) response to the high therapeutic pressure exerted by chemoradiation. We found EMT to be initiated by the autocrine production and response to transforming growth factor beta (TGF-β) of EAC cells. Inhibition of TGF-β ligands effectively abolished chemoradiation-induced EMT. Assessment of TGF-β serum levels in EAC patients revealed that high levels after neoadjuvant treatment predicted the presence of fluorodeoxyglucose uptake in lymph nodes on the post-chemoradiation positron emission tomography-scan. Our study shows that chemoradiation contributes to resistant metastatic disease in EAC patients by inducing EMT via autocrine TGF-β production. Monitoring TGF-β serum levels during treatment could identify those patients at risk of developing metastatic disease, and who would likely benefit from TGF-β targeting therapy.
Sigma-Aldrich
Monoclonal Anti-ZEB1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0151, purified immunoglobulin, buffered aqueous glycerol solution
Pricing and availability is not currently available.