Merck
  • Elevated prolactin secretion during proestrus in mice: Absence of a defined surge.
Elevated prolactin secretion during proestrus in mice: Absence of a defined surge.
Journal of neuroendocrinology (2022-05-02)
Hollian R Phillipps, Zin Khant Aung, David R Grattan
ABSTRACT
Throughout the reproductive cycle in rodents, prolactin levels are generally low. In some species, including rats, a prolactin surge occurs on proestrus with peak concentrations coinciding with the preovulatory luteinizing hormone (LH) surge. In mice, however, there are conflicting reports relating to the occurrence and timing of a proestrous prolactin surge. To gain further insight into the incidence and characteristics of this surge in mice, we have used serial tail tip blood sampling and trunk blood collection from both C57BL/6J (inbred) and Swiss Webster (outbred) mouse strains to build a profile of prolactin secretion during proestrus in individual mice. A clearly defined LH surge was detected in most animals, suggesting the blood sampling approach was suitable for detecting patterns of hormone secretion on proestrus. Despite this, levels of prolactin were quite variable between individuals. Overall both mouse strains showed a generalized rise in prolactin levels on the day of proestrus compared with levels seen in diestrus. This pattern is quite distinct from the discreet, circadian-entrained surge observed in rats.
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 20 mg substrate per tablet
Pricing and availability is not currently available.