Merck
  • Synthesis and in vitro evaluation of bisphosphonated glycopeptide prodrugs for the treatment of osteomyelitis.
Synthesis and in vitro evaluation of bisphosphonated glycopeptide prodrugs for the treatment of osteomyelitis.
Bioorganic & medicinal chemistry letters (2010-01-26)
Kelly S E Tanaka, Evelyne Dietrich, Stéphane Ciblat, Claude Métayer, Francis F Arhin, Ingrid Sarmiento, Gregory Moeck, Thomas R Parr, Adel Rafai Far
ABSTRACT
As therapeutic agents of choice in the treatment of complicated infections, glycopeptide antibiotics are often preferentially used in cases of osteomyelitis, an infection located in bone and notoriously difficult to successfully manage. Yet frequent and heavy doses of these systemically administered antibiotics are conventionally prescribed to obtain higher antibiotic levels in the bone and reduce the high recurrence rates. Targeting antibiotics to the bone after systemic administration would present at least three potential advantages: (i) greater efficacy, by concentrating the therapeutic agent in bone; (ii) greater convenience, through a reduction in the frequency of administration; and (iii) greater safety, by reducing the levels of systemic drug exposure. We present here the design, synthesis and in vitro evaluation of eight prodrugs of the glycopeptide antibacterial agents vancomycin and oritavancin taking advantage of the affinity of the bisphosphonate group for bone for delivery to osseous tissues.
Sigma-Aldrich
Methylenebis(phosphonic dichloride), 97%
Pricing and availability is not currently available.