Merck
  • MCL1 nuclear translocation induces chemoresistance in colorectal carcinoma.
MCL1 nuclear translocation induces chemoresistance in colorectal carcinoma.
Cell death & disease (2022-01-20)
Dechen Fu, Luke Pfannenstiel, Abeba Demelash, Yee Peng Phoon, Cameron Mayell, Claudia Cabrera, Caini Liu, Junjie Zhao, Josephine Dermawan, Deepa Patil, Jennifer DeVecchio, Matthew Kalady, Andrew J Souers, Darren C Phillips, Xiaoxia Li, Brian Gastman
ABSTRACT
Colorectal cancer (CRC) is one of the most common and deadliest forms of cancer. Myeloid Cell Leukemia 1 (MCL1), a pro-survival member of the Bcl-2 protein family is associated with chemo-resistance in CRC. The ability of MCL1 to inhibit apoptosis by binding to the BH3 domains of pro-apoptotic Bcl-2 family members is a well-studied means by which this protein confers resistance to multiple anti-cancer therapies. We found that specific DNA damaging chemotherapies promote nuclear MCL1 translocation in CRC models. In p53null CRC, this process is associated with resistance to chemotherapeutic agents, the mechanism of which is distinct from the classical mitochondrial protection. We previously reported that MCL1 has a noncanonical chemoresistance capability, which requires a novel loop domain that is distinct from the BH3-binding domain associated with anti-apoptotic function. Herein we disclose that upon treatment with specific DNA-damaging chemotherapy, this loop domain binds directly to alpha-enolase which in turn binds to calmodulin; we further show these protein-protein interactions are critical in MCL1's nuclear import and chemoresistance. We additionally observed that in chemotherapy-treated p53-/- CRC models, MCL1 nuclear translocation confers sensitivity to Bcl-xL inhibitors, which has significant translational relevance given the co-expression of these proteins in CRC patient samples. Together these findings indicate that chemotherapy-induced MCL1 translocation represents a novel resistance mechanism in CRC, while also exposing an inherent and targetable Bcl-xL co-dependency in these cancers. The combination of chemotherapy and Bcl-xL inhibitors may thus represent a rational means of treating p53-/- CRC via exploitation of this unique MCL1-based chemoresistance mechanism.
Sigma-Aldrich
Calmidazolium Chloride, A cell-permeable calmodulin antagonist.