Merck
  • UPLC-MS, HPLC-radiometric, and NMR-spectroscopic studies on the metabolic fate of 3-fluoro-[U-14C]-aniline in the bile-cannulated rat.
UPLC-MS, HPLC-radiometric, and NMR-spectroscopic studies on the metabolic fate of 3-fluoro-[U-14C]-aniline in the bile-cannulated rat.
Xenobiotica; the fate of foreign compounds in biological systems (2010-05-07)
T J Athersuch, J Castro-Perez, C Rodgers, J K Nicholson, Ian D Wilson
ABSTRACT
1. A study of the rates and routes of excretion of 3-fluoro-[U-(14)C]aniline following intraperitoneal administration to male bile-cannulated rats by liquid scintillation counting (LSC) gave a total recovery of approximately 90% in the 48 h following dosing, with the majority of the dose being excreted in the urine during the first 24 h (approximately 49%). 2. The total recovery as determined by (19)F-nuclear magnetic resonance ((19)F-NMR) was approximately 49%, with the majority of the dose excreted in the first 24 h (approximately 41%). The comparatively low recovery in comparison to that obtained from LSC was due to matrix effects in bile and a contribution from metabolic defluorination. 3. High-performance liquid chromatography with radiometric profiling of urine and bile revealed a complex pattern of metabolites with the bulk of the dose excreted as a single peak. 4. Ultra-performance liquid chromatography-orthogonal acceleration time of flight mass spectrometry profiling also showed a complex pattern of metabolites, detecting approximately 21 metabolites of 3-fluoroaniline (3-FA) with six of these detected only in urine and four solely in bile. 5. (19)F-NMR revealed the presence of the parent compound and 15 metabolites in urine collected during the first 24 h after -dosing. The matrix effects of bile on (19)F-NMR spectroscopy made metabolite profiling impractical for this biofluid. The major metabolite of 3-FA was identified as 2-fluoro-4-acetamidophenol-sulfate.
Sigma-Aldrich
Triton X-405 solution, 70% in H2O
Pricing and availability is not currently available.