Merck
  • Antiangiogenic and antitumor activity of a selective PDGFR tyrosine kinase inhibitor, CP-673,451.
Antiangiogenic and antitumor activity of a selective PDGFR tyrosine kinase inhibitor, CP-673,451.
Cancer research (2005-02-12)
W Gregory Roberts, Pamela M Whalen, Erik Soderstrom, Garrett Moraski, Joseph P Lyssikatos, Huifen-F Wang, Beth Cooper, Deborah A Baker, Douglas Savage, Deepak Dalvie, James A Atherton, Sherry Ralston, Ruby Szewc, John C Kath, Jing Lin, Cathy Soderstrom, George Tkalcevic, Bruce D Cohen, Vince Pollack, Wayne Barth, Will Hungerford, Ethan Ung
ABSTRACT
CP-673,451 is a potent inhibitor of platelet-derived growth factor beta-receptor (PDGFR-beta) kinase- and PDGF-BB-stimulated autophosphorylation of PDGFR-beta in cells (IC(50) = 1 nmol/L) being more than 450-fold selective for PDGFR-beta versus other angiogenic receptors (e.g., vascular endothelial growth factor receptor 2, TIE-2, and fibroblast growth factor receptor 2). Multiple models have been used to evaluate in vivo activity of CP-673,451 and to understand the pharmacology of PDGFR-beta inhibition and the effect on tumor growth. These models include an ex vivo measure of PDGFR-beta phosphorylation in glioblastoma tumors, a sponge model to measure inhibition of angiogenesis, and multiple models of tumor growth inhibition. Inhibition of PDGFR-beta phosphorylation in tumors correlates with plasma and tumor levels of CP-673,451. A dose of 33 mg/kg was adequate to provide >50% inhibition of receptor for 4 hours corresponding to an EC(50) of 120 ng/mL in plasma at C(max). In a sponge angiogenesis model, CP-673,451 inhibited 70% of PDGF-BB-stimulated angiogenesis at a dose of 3 mg/kg (q.d. x 5, p.o., corresponding to 5.5 ng/mL at C(max)). The compound did not inhibit vascular endothelial growth factor- or basic fibroblast growth factor-induced angiogenesis at concentrations which inhibited tumor growth. The antitumor efficacy of CP-673,451 was evaluated in a number of human tumor xenografts grown s.c. in athymic mice, including H460 human lung carcinoma, Colo205 and LS174T human colon carcinomas, and U87MG human glioblastoma multiforme. Once-daily p.o. x 10 days dosing routinely inhibited tumor growth (ED(50) < or = 33 mg/kg). These data show that CP-673,451 is a pharmacologically selective PDGFR inhibitor, inhibits tumor PDGFR-beta phosphorylation, selectively inhibits PDGF-BB-stimulated angiogenesis in vivo, and causes significant tumor growth inhibition in multiple human xenograft models.
Sigma-Aldrich
CP-673451, ≥95% (HPLC)
Pricing and availability is not currently available.