Merck
  • Distinct regulation of tonic GABAergic inhibition by NMDA receptor subtypes.
Distinct regulation of tonic GABAergic inhibition by NMDA receptor subtypes.
Cell reports (2021-11-11)
Kunwei Wu, David Castellano, Qingjun Tian, Wei Lu
ABSTRACT
Tonic inhibition mediated by extrasynaptic GABAARs regulates various brain functions. However, the mechanisms that regulate tonic inhibition remain largely unclear. Here, we report distinct actions of GluN2A- and GluN2B-NMDA receptors (NMDARs) on tonic inhibition in hippocampal neurons under basal and high activity conditions. Specifically, overexpression of GluN2B, but not GluN2A, reduces α5-GABAAR surface expression and tonic currents. Additionally, knockout of GluN2A and GluN2B decreases and increases tonic currents, respectively. Mechanistically, GluN2A-NMDARs inhibit and GluN2B-NMDARs promote α5-GABAAR internalization, resulting in increased and decreased surface α5-GABAAR expression, respectively. Furthermore, GluN2A-NMDARs, but not GluN2B-NMDARs, are required for homeostatic potentiation of tonic inhibition induced by prolonged increase of neuronal activity. Last, tonic inhibition decreases during acute seizures, whereas it increases 24 h later, involving GluN2-NMDAR-dependent signaling. Collectively, these data reveal an NMDAR subunit-specific regulation of tonic inhibition in physiological and pathological conditions and provide mechanistic insight into activity-dependent modulation of tonic inhibition.
Sigma-Aldrich
Anti-Glutamate Receptor NMDAR2B (NR2B) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Picrotoxin, powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-GABAA Receptor α1 Antibody, Upstate®, from rabbit
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Glutamate Receptor NMDAR2A (NR2A) antibody produced in rabbit, affinity isolated antibody, lyophilized powder