Merck
  • Caveolin-1 temporal modulation enhances antibody drug efficacy in heterogeneous gastric cancer.
Caveolin-1 temporal modulation enhances antibody drug efficacy in heterogeneous gastric cancer.
Nature communications (2022-05-10)
Patrícia M R Pereira, Komal Mandleywala, Sébastien Monette, Melissa Lumish, Kathryn M Tully, Sandeep Surendra Panikar, Mike Cornejo, Audrey Mauguen, Ashwin Ragupathi, Nai C Keltee, Marissa Mattar, Yelena Y Janjigian, Jason S Lewis
ABSTRACT
Resistance mechanisms and heterogeneity in HER2-positive gastric cancers (GC) limit Trastuzumab benefit in 32% of patients, and other targeted therapies have failed in clinical trials. Using patient samples, patient-derived xenografts (PDXs), partially humanized biological models, and HER2-targeted imaging technologies we demonstrate the role of caveolin-1 (CAV1) as a complementary biomarker in GC selection for Trastuzumab therapy. In retrospective analyses of samples from patients enrolled on Trastuzumab trials, the CAV1-high profile associates with low membrane HER2 density and low patient survival. We show a negative correlation between CAV1 tumoral protein levels - a major protein of cholesterol-rich membrane domains - and Trastuzumab-drug conjugate TDM1 tumor uptake. Finally, CAV1 depletion using knockdown or pharmacologic approaches (statins) increases antibody drug efficacy in tumors with incomplete HER2 membranous reactivity. In support of these findings, background statin use in patients associates with enhanced antibody efficacy. Together, this work provides preclinical justification and clinical evidence that require prospective investigation of antibody drugs combined with statins to delay drug resistance in tumors.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse