Merck
  • The emerging role of xanthine oxidase inhibition for suppression of breast cancer cell migration and metastasis associated with hypercholesterolemia.
The emerging role of xanthine oxidase inhibition for suppression of breast cancer cell migration and metastasis associated with hypercholesterolemia.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2019-03-13)
Se-Hyun Oh, Soon-Youn Choi, Hyo-Jung Choi, Hye-Myung Ryu, You-Jin Kim, Hee-Yeon Jung, Jang-Hee Cho, Chan-Duck Kim, Sun-Hee Park, Tae-Hwan Kwon, Yong-Lim Kim
ABSTRACT
Hypercholesterolemia is reported to increase reactive oxygen species (ROS) and to promote breast cancer progression. ROS play an important role in tumor biology, and xanthine oxidase (XO) is an enzyme that generates ROS. The effects of febuxostat (FBX), an XO inhibitor, on breast cancer cell migration under LDL stimulation in vitro and metastasis of breast cancer associated with hypercholesterolemia in vivo were studied. In vitro, FBX significantly inhibited LDL-induced ROS production and cell migration. Treatment of small interfering RNA against XO was consistent with the findings of FBX treatment. In vivo, a significant increase of tumor growth and pulmonary metastasis was observed in a xenograft mouse model with 4T1 cells on a high cholesterol diet (HCD), both of which were markedly inhibited by FBX or allopurinol treatment. Moreover, ERK represented the main target-signaling pathway that was affected by FBX treatment in a xenograft mouse model on an HCD evaluated by NanoString nCounter analysis. Consistently, MEK/ERK inhibitors directly decreased the LDL-induced cell migration in vitro. In conclusion, FBX mitigates breast cancer cell migration and pulmonary metastasis in the hyperlipidemic condition, associated with decreased ROS generation and MAPK phosphorylation. The inhibition of ERK pathways is likely to underlie the XO inhibitor-mediated suppression of breast cancer cell migration.-Oh, S.-H., Choi, S.-Y., Choi, H.-J., Ryu, H.-M., Kim, Y.-J., Jung, H.-Y., Cho, J.-H., Kim, C.-D., Park, S.-H., Kwon, T.-H., Kim, Y.-L. The emerging role of xanthine oxidase inhibition for suppression of breast cancer cell migration and metastasis associated with hypercholesterolemia.
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Lipoproteins, Low Density, Human Plasma
Pricing and availability is not currently available.