Merck
  • Differential regulation of actin-activated nucleotidyl cyclase virulence factors by filamentous and globular actin.
Differential regulation of actin-activated nucleotidyl cyclase virulence factors by filamentous and globular actin.
PloS one (2018-11-13)
Dorothée Raoux-Barbot, Alexander Belyy, Lina Worpenberg, Sabrina Montluc, Celia Deville, Véronique Henriot, Christophe Velours, Daniel Ladant, Louis Renault, Undine Mechold
ABSTRACT
Several bacterial pathogens produce nucleotidyl cyclase toxins to manipulate eukaryotic host cells. Inside host cells they are activated by endogenous cofactors to produce high levels of cyclic nucleotides (cNMPs). The ExoY toxin from Pseudomonas aeruginosa (PaExoY) and the ExoY-like module (VnExoY) found in the MARTX (Multifunctional-Autoprocessing Repeats-in-ToXin) toxin of Vibrio nigripulchritudo share modest sequence similarity (~38%) but were both recently shown to be activated by actin after their delivery to the eukaryotic host cell. Here, we further characterized the ExoY-like cyclase of V. nigripulchritudo. We show that, in contrast to PaExoY that requires polymerized actin (F-actin) for maximum activation, VnExoY is selectively activated by monomeric actin (G-actin). These two enzymes also display different nucleotide substrate and divalent cation specificities. In vitro in presence of the cation Mg2+, the F-actin activated PaExoY exhibits a promiscuous nucleotidyl cyclase activity with the substrate preference GTP>ATP≥UTP>CTP, while the G-actin activated VnExoY shows a strong preference for ATP as substrate, as it is the case for the well-known calmodulin-activated adenylate cyclase toxins from Bordetella pertussis or Bacillus anthracis. These results suggest that the actin-activated nucleotidyl cyclase virulence factors despite sharing a common activator may actually display a greater variability of biological effects in infected cells than initially anticipated.