- Pyruvate Kinase M1 Suppresses Development and Progression of Prostate Adenocarcinoma.
Pyruvate Kinase M1 Suppresses Development and Progression of Prostate Adenocarcinoma.
Cancer research (2022-05-19)
Shawn M Davidson, Daniel R Schmidt, Julia E Heyman, James P O'Brien, Amy C Liu, William J Israelsen, Talya L Dayton, Raghav Sehgal, Roderick T Bronson, Elizaveta Freinkman, Howard H Mak, Giuseppe Nicolò Fanelli, Scott Malstrom, Gary Bellinger, Arkaitz Carracedo, Pier Paolo Pandolfi, Kevin D Courtney, Abhishek Jha, Ronald A DePinho, James W Horner, Craig J Thomas, Lewis C Cantley, Massimo Loda, Matthew G Vander Heiden
PMID35584006
ABSTRACT
Differential expression of PKM1 and PKM2 impacts prostate tumorigenesis and suggests a potential therapeutic vulnerability in prostate cancer.
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone VIN-11-5, ascites fluid
Sigma-Aldrich
Anti-PKM2 (isoform M1) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody