Merck
  • Danger signal extracellular calcium initiates differentiation of monocytes into SPP1/osteopontin-producing macrophages.
Danger signal extracellular calcium initiates differentiation of monocytes into SPP1/osteopontin-producing macrophages.
Cell death & disease (2022-01-14)
Supriya Murthy, Isabel Karkossa, Caroline Schmidt, Anne Hoffmann, Tobias Hagemann, Kathrin Rothe, Olga Seifert, Ulf Anderegg, Martin von Bergen, Kristin Schubert, Manuela Rossol
ABSTRACT
The danger signal extracellular calcium is pathophysiologically increased in the synovial fluid of patients with rheumatoid arthritis (RA). Calcium activates the NLRP3-inflammasome via the calcium-sensing receptor in monocytes/macrophages primed by lipopolysaccharide, and this effect is mediated by the uptake of calciprotein particles (CPPs) formed out of calcium, phosphate, and fetuin-A. Aim of the study was to unravel the influence of calcium on monocytes when the priming signal is not present. Monocytes were isolated from the blood of healthy controls and RA patients. Macrophages were characterized using scRNA-seq, DNA microarray, and proteomics. Imaging flow cytometry was utilized to study intracellular events. Here we show that extracellular calcium and CPPs lead to the differentiation of monocytes into calcium-macrophages when the priming signal is absent. Additional growth factors are not needed, and differentiation is triggered by calcium-dependent CPP-uptake, lysosomal alkalization due to CPP overload, and TFEB- and STAT3-dependent increased transcription of the lysosomal gene network. Calcium-macrophages have a needle-like shape, are characterized by excessive, constitutive SPP1/osteopontin production and a strong pro-inflammatory cytokine response. Calcium-macrophages differentiated out of RA monocytes show a stronger manifestation of this phenotype, suggesting the differentiation process might lead to the pro-inflammatory macrophage response seen in the RA synovial membrane.
Sigma-Aldrich
STAT3 Inhibitor VI, S3I-201, STAT3 Inhibitor VI, S3I-201, CAS 501919-59-1, is a cell-permeable compound that binds Stat3-SH2 domain and blocks Stat3 phosphorylation, dimerization, DNA-binding, & Stat3-dependent transcription.
Pricing and availability is not currently available.