Merck
  • Novel GLP-1/GLP-2 co-agonists display marked effects on gut volume and improves glycemic control in mice.
Novel GLP-1/GLP-2 co-agonists display marked effects on gut volume and improves glycemic control in mice.
Physiology & behavior (2018-03-16)
Pernille Wismann, Søren L Pedersen, Gitte Hansen, Karin Mannerstedt, Philip J Pedersen, Palle B Jeppesen, Niels Vrang, Keld Fosgerau, Jacob Jelsing
ABSTRACT
Analogues of several gastrointestinal peptide hormones have been developed into effective medicines for treatment of diseases such as type 2 diabetes mellitus (T2DM), obesity and short bowel syndrome (SBS). In this study, we aimed to explore whether the combination of glucagon-like peptide-1 (GLP-1) and glucagon-like peptide-2 (GLP-2) into a potent co-agonist could provide additional benefits compared to existing monotherapies. A short-acting (GUB09-123) and a half-life extended (GUB09-145) GLP-1/GLP-2 co-agonist were generated using solid-phase peptide synthesis and tested for effects on food intake, body weight, glucose homeostasis, and gut proliferation in lean mice and in diabetic db/db mice. Sub-chronic administration of GUB09-123 to lean mice significantly reduced food intake, improved glucose tolerance, and increased gut volume, superior to monotherapy with the GLP-2 analogue teduglutide. Chronic administration of GUB09-123 to diabetic mice significantly improved glycemic control and showed persistent effects on gastric emptying, superior to monotherapy with the GLP-1 analogue liraglutide. Due to the short-acting nature of the molecule, no effects on body weight were observed, whereas a marked and robust intestinotrophic effect on mainly the small intestine volume and surface area was obtained. In contrast to GUB09-123, sub-chronic administration of a half-life extended GUB09-145 to lean mice caused marked dose-dependent effects on body weight while maintaining its potent intestinotrophic effect. Our data demonstrate that the GLP-1/GLP-2 co-agonists have effects on gut morphometry, showing a marked increase in intestinal volume and mucosal surface area. Furthermore, effects on glucose tolerance and long-term glycemic control are evident. Effects on body weight and gastric emptying are also observed depending on the pharmacokinetic properties of the molecule. We suggest that this novel co-agonistic approach could exemplify a novel concept for treatment of T2DM or SBS.
Sigma-Aldrich
Teduglutide trifluoroacetate, ≥95% (HPLC)