Merck
  • Prolyl hydroxylase inhibitor desidustat improves anemia in erythropoietin hyporesponsive state.
Prolyl hydroxylase inhibitor desidustat improves anemia in erythropoietin hyporesponsive state.
Current research in pharmacology and drug discovery (2022-05-17)
Amit A Joharapurkar, Vishal J Patel, Samadhan G Kshirsagar, Maulik S Patel, Hardikkumar H Savsani, Chetan Kajavadara, Darshan Valani, Mukul R Jain
ABSTRACT
Many anemic chronic kidney disease (CKD) patients are refractory to erythropoietin (EPO) effects due to inflammation, deranged iron utilization, and generation of EPO antibodies. This work assessed the effect of desidustat, an inhibitor of hypoxia inducible factor (HIF) prolyl hydroxylase (PHD), on EPO-refractory renal anemia. Sprague Dawley rats were made anemic by cisplatin (5 ​mg/kg, IP, single dose) and turpentine oil (5 ​mL/kg, SC, once a week). These rats were given recombinant human EPO (rhEPO, 1 ​μg/kg) and desidustat (15 or 30 ​mg/kg) for eight weeks. Separately, rhEPO (1-5 ​μg/kg) was given to anemic rats to sustain the normal hemoglobin levels and desidustat (15 ​mg/kg) for eight weeks. In another experiment, the anemic rats were treated rhEPO (5 ​μg/kg) for two weeks and then desidustat (15 ​mg/kg) for the next two weeks. Dosing of rhEPO was thrice a week, and for desidustat, it was on alternate days. Desidustat inhibited EPO-resistance caused by rhEPO treatment, decreased hepcidin, IL-6, IL-1β, and increased iron and liver ferroportin. Desidustat reduced EPO requirement and anti-EPO antibodies. Desidustat also maintained normal hemoglobin levels after cessation of rhEPO treatment. Thus, novel prolyl hydroxylase inhibitor desidustat can treat EPO resistance via improved iron utilization and decreased inflammation.
Sigma-Aldrich
Anti-Erythropoietin antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
Pricing and availability is not currently available.