Merck
  • Facilitating understanding of the purine nucleotide cycle and the one-carbon pool: Part I: The purine nucleotide cycle.
Facilitating understanding of the purine nucleotide cycle and the one-carbon pool: Part I: The purine nucleotide cycle.
Biochemistry and molecular biology education : a bimonthly publication of the International Union of Biochemistry and Molecular Biology (2005-05-01)
Ifeanyi J Arinze
ABSTRACT
Some metabolic processes are readily understood because they are circumscribed in metabolic pathways that have clearly identifiable beginning points, end products, and other features. Other metabolic pathways that do not appear to be straightforward pose difficulties for students. One such metabolic process, the purine nucleotide cycle, is discussed here.
Sigma-Aldrich
Aspartate Metabolite Library