Merck
Fiber composite slices for multiplexed immunoassays.
Biomicrofluidics (2015-09-05)
Jiyun Kim, Sangwook Bae, Seowoo Song, Keumsim Chung, Sunghoon Kwon
ABSTRACT
Fabrication methods for the development of multiplexed immunoassay platforms primarily depend on the individual functionalization of reaction chambers to achieve a heterogeneous reacting substrate composition, which increases the overall manufacturing time and cost. Here, we describe a new type of low-cost fabrication method for a scalable immunoassay platform based on cotton threads. The manufacturing process involves the fabrication of functionalized fibers and the arrangement of these fibers into a bundle; this bundle is then sectioned to make microarray-like particles with a predefined surface architecture. With these sections, composed of heterogeneous thread fragments with different types of antibodies, we demonstrated quantitative and 7-plex immunoassays. We expect that this methodology will prove to be a versatile, low-cost, and highly scalable method for the fabrication of multiplexed bioassay platforms.
Sigma-Aldrich
Interleukin-12 Protein, Recombinant human, The recombinant human Interleukin-12 protein (or IL-12 protein) is a potent regulator of cell mediated immune responses & it induces IFN-gamma production by NK & T cells.
Pricing and availability is not currently available.