Merck
  • Characterization of MS/MS Product Ions for the Differentiation of Structurally Isomeric Pesticides by High-Resolution Mass Spectrometry.
Characterization of MS/MS Product Ions for the Differentiation of Structurally Isomeric Pesticides by High-Resolution Mass Spectrometry.
Toxics (2018-10-04)
Alberto Nuñez, Yelena Sapozhnikova, Steven J Lehotay
ABSTRACT
Structural isomeric pesticides are used in agriculture and may be challenging to differentiate for accurate identification in pesticide monitoring programs. Due to structural similarity, isomeric pesticides are difficult to separate chromatographically, and thus, their accurate identification may rely solely on mass spectrometric analysis (MS). In this study, we challenged the ability of high-resolution quadrupole-orbitrap (Q-Orbitrap) mass spectrometry to produce and evaluate the tandem mass spectrometry (MS/MS) product ions for the selected five pairs of isomeric pesticides from different classes: Pebulate and vernolate, methiocarb and ethiofencarb, uniconazole and cyproconazole, sebuthylazine and terbuthylazine, and orbencarb and thiobencarb. The use of Q-Orbitrap instrument with a mass error <3 ppm allowed proposed elucidation of the product ion structures with consideration of the ion formulae, data interpretation, and literature searches. Product ions unique to pebulate, vernolate, methiocarb, ethiofencarb, and uniconazole were observed. Elucidation of the observed MS/MS product ion structures was conducted, and the fragmentation pathways were proposed. This information is valuable to increase selectivity in MS/MS analysis and differentiate isomeric pesticides, and thereby reduce the rates of false positives in pesticide monitoring programs.
Product Description
Supelco
Ethiofencarb, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.