Merck
  • Water extract of propolis and its main constituents, caffeoylquinic acid derivatives, exert neuroprotective effects via antioxidant actions.
Water extract of propolis and its main constituents, caffeoylquinic acid derivatives, exert neuroprotective effects via antioxidant actions.
Life sciences (2006-10-19)
Yoshimi Nakajima, Masamitsu Shimazawa, Satoshi Mishima, Hideaki Hara
ABSTRACT
We investigated whether water extract of Brazilian green propolis (WEP) and its main constituents [caffeoylquinic acid derivatives (3,4-di-O-caffeoylquinic acid, 3,5-di-O-caffeoylquinic acid, chlorogenic acid) and cinnamic acid derivatives (p-coumaric acid, artepillin C, drupanin, baccharin)] exert neuroprotective effects against the retinal damage induced by oxidative stress. Additionally, their neuroprotective effects were compared with their antioxidant effects. WEP, 3,4-di-O-caffeoylquinic acid, 3,5-di-O-caffeoylquinic acid, chlorogenic acid, and p-coumaric acid (but not artepillin C, baccharin, or drupanin) concentration-dependently inhibited oxidative stress-induced neurotoxicity [achieved using L-buthionine-(S,R)-sulfoximine (BSO) to deplete glutathione in combination with glutamate to inhibit cystine uptake] in cultured retinal ganglion cells (RGC-5, a rat ganglion cell line transformed using E1A virus). At their effective concentrations against oxidative stress-induced retinal damage, WEP, 3,4-di-caffeoylquinic acid, 3,5-di-caffeoylquinic acid, and chlorogenic acid (but not cinnamic acid derivatives) inhibited lipid peroxidation (LPO) in mouse forebrain homogenates. Thus, the neuroprotective effects of WEP and caffeoylquinic acid derivatives paralleled those against LPO. These findings indicate that WEP and caffeoylquinic acid derivatives have neuroprotective effects against retinal damage in vitro, and that these effects may be partly mediated via antioxidant effects.
Sigma-Aldrich
3,4-Di-O-caffeoylquinic acid, ≥90% (LC/MS-ELSD)