Merck
  • Synthesis and characterization of a new fluorogenic substrate for alpha-galactosidase.
Synthesis and characterization of a new fluorogenic substrate for alpha-galactosidase.
Analytical and bioanalytical chemistry (2009-06-13)
Zhen-Dan Shi, Omid Motabar, Ehud Goldin, Ke Liu, Noel Southall, Ellen Sidransky, Christopher P Austin, Gary L Griffiths, Wei Zheng
ABSTRACT
Alpha-galactosidase A hydrolyzes the terminal alpha-galactosyl moieties from glycolipids and glycoproteins in lysosomes. Mutations in alpha-galactosidase cause lysosomal accumulation of the glycosphingolipid, globotriaosylceramide, which leads to Fabry disease. Small-molecule chaperones that bind to mutant enzyme proteins and correct their misfolding and mistrafficking have emerged as a potential therapy for Fabry disease. We have synthesized a red fluorogenic substrate, resorufinyl alpha-D-galactopyranoside, for a new alpha-galactosidase enzyme assay. This assay can be measured continuously at lower pH values, without the addition of a stop solution, due to the relatively low pK(a) of resorufin (approximately 6). In addition, the assay emits red fluorescence, which can significantly reduce interferences due to compound fluorescence and dust/lint as compared to blue fluorescence. Therefore, this new red fluorogenic substrate and the resulting enzyme assay can be used in high-throughput screening to identify small-molecule chaperones for Fabry disease.
Sigma-Aldrich
4-Methylumbelliferyl α-D-galactopyranoside, ≥98% (TLC)
Pricing and availability is not currently available.