Merck
  • Selective Utilization of the Human Milk Oligosaccharides 2'-Fucosyllactose, 3-Fucosyllactose, and Difucosyllactose by Various Probiotic and Pathogenic Bacteria.
Selective Utilization of the Human Milk Oligosaccharides 2'-Fucosyllactose, 3-Fucosyllactose, and Difucosyllactose by Various Probiotic and Pathogenic Bacteria.
Journal of agricultural and food chemistry (2021-01-01)
Krista Salli, Johanna Hirvonen, Jani Siitonen, Ilmari Ahonen, Heli Anglenius, Johanna Maukonen
ABSTRACT
Prebiotic human milk oligosaccharides (HMOs) are found in human milk, which are not digested by infants but are metabolized by beneficial gut bacteria. We determined the ability of 57 bacterial strains within the Family Lactobacillaceae and genera Bifidobacterium and Bacteroides and potentially pathogenic bacteria to ferment the HMOs 2'-fucosyllactose, 3-fucosyllactose, and difucosyllactose. In addition, prebiotic galacto-oligosaccharides (GOS), lactose, fucose, and glucose were evaluated as carbon sources for these bacterial strains. Bacterial growth was monitored using the automatic Bioscreen C system. Only certain bifidobacteria, such as Bifidobacterium longum subsp. infantis and Bifidobacterium bifidum, as well as Bacteroides fragilis, Bacteroides vulgatus, and Bacteroides thetaiotaomicron utilized the studied HMOs as their sole carbon source, whereas almost all studied bacterial strains were able to utilize GOS, lactose, and glucose. The selectivity in utilization of HMOs by only certain bacteria can be advantageous by promoting beneficial microbes but not supporting the harmful pathogens in contrast to other less selective prebiotics.
Sigma-Aldrich
3-Fucosyllactose, ≥95% (HPLC)