Merck
  • Stereoselective synthesis of β-alkylated α-amino acids via palladium-catalyzed alkylation of unactivated methylene C(sp3)-H bonds with primary alkyl halides.
Stereoselective synthesis of β-alkylated α-amino acids via palladium-catalyzed alkylation of unactivated methylene C(sp3)-H bonds with primary alkyl halides.
Journal of the American Chemical Society (2013-08-08)
Shu-Yu Zhang, Qiong Li, Gang He, William A Nack, Gong Chen
ABSTRACT
We report a new set of reactions based on the Pd-catalyzed alkylation of methylene C(sp(3))-H bonds of aliphatic quinolyl carboxamides with α-haloacetate and methyl iodide and applications in the stereoselective synthesis of various β-alkylated α-amino acids. These reactions represent the first generally applicable method for the catalytic alkylation of unconstrained and unactivated methylene C-H bonds with high synthetic relevance. When applied with simple isotope-enriched reagents, they also provide a convenient and powerful means to site-selectively incorporate isotopes into the carbon scaffolds of amino acid compounds.