Merck
  • Recapitulating hepatitis E virus-host interactions and facilitating antiviral drug discovery in human liver-derived organoids.
Recapitulating hepatitis E virus-host interactions and facilitating antiviral drug discovery in human liver-derived organoids.
Science advances (2022-01-20)
Pengfei Li, Yunlong Li, Yijin Wang, Jiaye Liu, Marla Lavrijsen, Yang Li, Ruyi Zhang, Monique M A Verstegen, Yining Wang, Tian-Cheng Li, Zhongren Ma, Denis E Kainov, Marco J Bruno, Robert A de Man, Luc J W van der Laan, Maikel P Peppelenbosch, Qiuwei Pan
ABSTRACT
Hepatotropic viruses naturally have narrow host and tissue tropisms, challenging the development of robust experimental models. The advent of organoid technology provides a unique opportunity for moving the field forward. Here, we demonstrate that three-dimensional cultured organoids from fetal and adult human liver with cholangiocyte or hepatocyte phenotype support hepatitis E virus (HEV) replication. Inoculation with infectious HEV particles demonstrates that human liver–derived organoids support the full life cycle of HEV infection. By directing organoids toward polarized monolayers in a transwell system, we observed predominantly apical secretion of HEV particles. Genome-wide transcriptomic and tRNAome analyses revealed robust host responses triggered by viral replication. Drug screening in organoids identified brequinar and homoharringtonine as potent HEV inhibitors, which are also effective against the ribavirin resistance variant harboring G1634R mutation. Thus, successful recapitulation of HEV infection in liver-derived organoids shall facilitate the study of virus-host interactions and development of antiviral therapies.