Merck
  • Mechanism of Cellular Formation and In Vivo Seeding Effects of Hexameric β-Amyloid Assemblies.
Mechanism of Cellular Formation and In Vivo Seeding Effects of Hexameric β-Amyloid Assemblies.
Molecular neurobiology (2021-10-06)
Céline Vrancx, Devkee M Vadukul, Nuria Suelves, Sabrina Contino, Ludovic D'Auria, Florian Perrin, Vincent van Pesch, Bernard Hanseeuw, Loïc Quinton, Pascal Kienlen-Campard
ABSTRACT
The β-amyloid peptide (Aβ) is found as amyloid fibrils in senile plaques, a typical hallmark of Alzheimer's disease (AD). However, intermediate soluble oligomers of Aβ are now recognized as initiators of the pathogenic cascade leading to AD. Studies using recombinant Aβ have shown that hexameric Aβ in particular acts as a critical nucleus for Aβ self-assembly. We recently isolated hexameric Aβ assemblies from a cellular model, and demonstrated their ability to enhance Aβ aggregation in vitro. Here, we report the presence of similar hexameric-like Aβ assemblies across several cellular models, including neuronal-like cell lines. In order to better understand how they are produced in a cellular context, we investigated the role of presenilin-1 (PS1) and presenilin-2 (PS2) in their formation. PS1 and PS2 are the catalytic subunits of the γ-secretase complex that generates Aβ. Using CRISPR-Cas9 to knockdown each of the two presenilins in neuronal-like cell lines, we observed a direct link between the PS2-dependent processing pathway and the release of hexameric-like Aβ assemblies in extracellular vesicles. Further, we assessed the contribution of hexameric Aβ to the development of amyloid pathology. We report the early presence of hexameric-like Aβ assemblies in both transgenic mice brains exhibiting human Aβ pathology and in the cerebrospinal fluid of AD patients, suggesting hexameric Aβ as a potential early AD biomarker. Finally, cell-derived hexameric Aβ was found to seed other human Aβ forms, resulting in the aggravation of amyloid deposition in vivo and neuronal toxicity in vitro.
Sigma-Aldrich
Anti-Amyloid β Antibody, clone W0-2, clone WO2, from mouse
Sigma-Aldrich
Anti-Amyloid β (Aβ)x-40 Antibody, clone 11A5-B10, ascites fluid, clone 11A5-B10, Upstate®
Sigma-Aldrich
Anti-Amyloid Beta (ABeta) x-42 Antibody, clone 12F4, clone 12F4, from mouse
Sigma-Aldrich
Anti-Amyloid Precursor Protein, C-Terminal antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution