Merck
  • Antisense Oligonucleotide-Mediated Reduction of HDAC6 Does Not Reduce Tau Pathology in P301S Tau Transgenic Mice.
Antisense Oligonucleotide-Mediated Reduction of HDAC6 Does Not Reduce Tau Pathology in P301S Tau Transgenic Mice.
Frontiers in neurology (2021-07-16)
Antonio Valencia, Veronica L Reinhart Bieber, Bekim Bajrami, Galina Marsh, Stefan Hamann, Ru Wei, Karen Ling, Frank Rigo, H Moore Arnold, Olga Golonzhka, Heike Hering
ABSTRACT
Acetylation of tau protein is dysregulated in Alzheimer's Disease (AD). It has been proposed that acetylation of specific sites in the KXGS motif of tau can regulate phosphorylation of nearby residues and reduce the propensity of tau to aggregate. Histone deacetylase 6 (HDAC6) is a cytoplasmic enzyme involved in deacetylation of multiple targets, including tau, and it has been suggested that inhibition of HDAC6 would increase tau acetylation at the KXGS motifs and thus may present a viable therapeutic approach to treat AD. To directly test the contribution of HDAC6 to tau pathology, we intracerebroventricularly injected an antisense oligonucleotide (ASO) directed against HDAC6 mRNA into brains of P301S tau mice (PS19 model), which resulted in a 70% knockdown of HDAC6 protein in the brain. Despite a robust decrease in levels of HDAC6, no increase in tau acetylation was observed. Additionally, no change of tau phosphorylation or tau aggregation was detected upon the knockdown of HDAC6. We conclude that HDAC6 does not impact tau pathology in PS19 mice.
Sigma-Aldrich
Anti-Cortactin (p80/85) Antibody, clone 4F11, clone 4F11, Upstate®, from mouse
Sigma-Aldrich
Anti-acetyl Cortactin Antibody, from rabbit, purified by affinity chromatography