Merck
  • The effect of continuous culture on the growth and structure of tissue-engineered cartilage.
The effect of continuous culture on the growth and structure of tissue-engineered cartilage.
Biotechnology progress (2009-03-19)
Aasma A Khan, Jocelyne M T Suits, Rita A Kandel, Stephen D Waldman
ABSTRACT
The use of bioreactors for cartilage tissue engineering has become increasingly important as traditional batch-fed culture is not optimal for in vitro tissue growth. Most tissue engineering bioreactors rely on convection as the primary means to provide mass transfer; however, convective transport can also impart potentially unwanted and/or uncontrollable mechanical stimuli to the cells resident in the construct. The reliance on diffusive transport may not necessarily be ineffectual as previous studies have observed improved cartilaginous tissue growth when the constructs were cultured in elevated volumes of media. In this study, to approximate an infinite reservoir of media, we investigated the effect of continuous culture on cartilaginous tissue growth in vitro. Isolated bovine articular chondrocytes were seeded in high density, 3D culture on Millicell filters. After two weeks of preculture, the constructs were cultivated with or without continuous media flow (5-10 microL/min) for a period of one week. Tissue engineered cartilage constructs grown under continuous media flow significantly accumulated more collagen and proteoglycans (increased by 50-70%). These changes were similar in magnitude to the reported effect of through-thickness perfusion without the need for large volumetric flow rates (5-10microL/min as opposed to 240-800 microL/min). Additionally, tissues grown in the reactor displayed some evidence of the stratified morphology of native cartilage as well as containing stores of intracellular glycogen. Future studies will investigate the effect of long-term continuous culture in terms of extracellular matrix accumulation and subsequent changes in mechanical function.