Merck
  • Dietary replacement of fish meal by defatted and fermented soybean meals with taurine supplementation for pompano fish: effects on growth performance, nutrient digestibility, and biological parameters in a long-term feeding period.
Dietary replacement of fish meal by defatted and fermented soybean meals with taurine supplementation for pompano fish: effects on growth performance, nutrient digestibility, and biological parameters in a long-term feeding period.
Journal of animal science (2020-11-18)
Hung Phuc Nguyen, Thinh Van Do, Hau Duc Tran
ABSTRACT
A 16-wk growth trial was conducted to examine the effects of dietary replacement of fish meal by defatted soybean meal (SBM) and fermented soybean meal (FSBM) with taurine supplementation on growth performance, nutrient apparent digestibility coefficient (ADC) and biological parameters of pompano fish. The FSBM was produced by fermenting SBM with Lactobacillus spp. Seven isonitrogenous and isoenergetic diets were formulated to replace 35% or 50% of fish meal by SBM or FSBM with taurine supplementation. The diets are denoted as follows: FM, SBM35, SBM35T, FSBM35T, SBM50, SBM50T, and FSBM50T. The FM (the basal diet) contained fish meal as a main source of dietary protein. Taurine was supplemented to SBM35T, FSBM35T, SBM50T, and FSBM50T at the level of 15 g/kg diet. Pompano juveniles with an initial body weight (BW) of 80 g reared in floating net cages were fed the experimental diets twice daily for 16 wk. Results showed that the final BW, weight gain, and feed conversion ratio of fish fed SBM35 and SBM50 were significantly lower than those of fish fed FM (P < 0.05), indicating that the replacement of fish meal by SBM at the rate of 35% in the diet is excessive for pompano. Supplementation of taurine to the SBM-included diets significantly increased growth performance and feed utilization (P < 0.05); however, these diets did not restore the performance back to a level equivalent to that of fish offered the basal diet. Meanwhile, fish fed FSBM35T had comparable growth and feed performances to those fed FM. Hematocrit values, total biliary bile acid levels, whole body lipid contents, and tissue taurine concentrations of fish fed SBM35 and SBM50 were the lowest among the treatments, but these parameters were improved by taurine supplementation and FSBM inclusion in the diet. Taurine supplementation increased lipid ADC, and SBM fermentation slightly enhanced both lipid and protein ADCs of the fish. These findings suggest that the combination of FSBM and taurine supplementation is an effective way to improve growth performance, nutrient digestibility, and biological parameters, and that FSBM with taurine supplementation can replace 35% of fish meal in pompano diets without any negative effects on growth and feed performances in a long-term feeding period.
Sigma-Aldrich
Bile Acid Assay Kit, sufficient for 100 fluorometric tests