Merck
  • Unveiling Extreme Photoreduction Potentials of Donor-Acceptor Cyanoarenes to Access Aryl Radicals from Aryl Chlorides.
Unveiling Extreme Photoreduction Potentials of Donor-Acceptor Cyanoarenes to Access Aryl Radicals from Aryl Chlorides.
Journal of the American Chemical Society (2021-08-26)
Jinhui Xu, Jilei Cao, Xiangyang Wu, Han Wang, Xiaona Yang, Xinxin Tang, Ren Wei Toh, Rong Zhou, Edwin K L Yeow, Jie Wu
ABSTRACT
Since the seminal work of Zhang in 2016, donor-acceptor cyanoarene-based fluorophores, such as 1,2,3,5-tetrakis(carbazol-9-yl)-4,6-dicyanobenzene (4CzIPN), have been widely applied in photoredox catalysis and used as excellent metal-free alternatives to noble metal Ir- and Ru-based photocatalysts. However, all the reported photoredox reactions involving this chromophore family are based on harnessing the energy from a single visible light photon, with a limited range of redox potentials from -1.92 to +1.79 V vs SCE. Here, we document the unprecedented discovery that this family of fluorophores can undergo consecutive photoinduced electron transfer (ConPET) to achieve very high reduction potentials. One of the newly synthesized catalysts, 2,4,5-tri(9H-carbazol-9-yl)-6-(ethyl(phenyl)amino)isophthalonitrile (3CzEPAIPN), possesses a long-lived (12.95 ns) excited radical anion form, 3CzEPAIPN•-*, which can be used to activate reductively recalcitrant aryl chlorides (Ered ≈ -1.9 to -2.9 V vs SCE) under mild conditions. The resultant aryl radicals can be engaged in synthetically valuable aromatic C-B, C-P, and C-C bond formation to furnish arylboronates, arylphosphonium salts, arylphosphonates, and spirocyclic cyclohexadienes.
Sigma-Aldrich
3CzEPAIPN, ≥95%
Pricing and availability is not currently available.