Merck
  • Cefcapene Pivoxil Hydrochloride Is a Potentially New Treatment for Palmoplantar Pustulosis with Pustulotic Arthro-Osteitis.
Cefcapene Pivoxil Hydrochloride Is a Potentially New Treatment for Palmoplantar Pustulosis with Pustulotic Arthro-Osteitis.
Dermatology (Basel, Switzerland) (2015-10-07)
Masamoto Murakami, Kana Masuda, Ryo Utsunomiya, Fumiko Oda, Chika Namba, Koji Sayama
ABSTRACT
Pustulosis palmaris et plantaris or palmoplantar pustulosis (PPP) is a refractory pustular eruption of the palms and soles with unknown etiology. In addition to skin lesions, PPP patients may present with severe joint pain and pustulotic arthro-osteitis (PAO), especially of the sternoclavicular joint. PAO is sometimes regarded as a variant of synovitis, acne, pustulosis, hyperostosis and osteitis (SAPHO) syndrome. Hence, macrolide and tetracycline antibiotics are used for the treatment of PPP with PAO. We report 3 cases of PPP with PAO that did not improve upon administration of macrolide antibiotics with NSAIDs. After administration of cefcapene pivoxil hydrochloride (CFPN-PI), a third-generation cephalosporin, the swelling and sternoclavicular joint pain were promptly reduced and dramatically improved in all 3 cases. We review the conventional antibiotic treatments used currently and propose CFPN-PI as a potentially new therapy for PPP or PPP + PAO.
Sigma-Aldrich
Cefcapene Pivoxil Hydrochloride Hydrate, ≥98% (HPLC)
Pricing and availability is not currently available.