Merck
  • Cortexolone 17α-Propionate (Clascoterone) is an Androgen Receptor Antagonist in Dermal Papilla Cells In Vitro
Cortexolone 17α-Propionate (Clascoterone) is an Androgen Receptor Antagonist in Dermal Papilla Cells In Vitro
Journal of drugs in dermatology : JDD (2019-02-28)
Caridad Rosette, Niccolette Rosette, Alessandro Mazzetti, Luigi Moro, Mara Gerloni
ABSTRACT
Cortexolone 17α-propionate (clascoterone) is a novel androgen antagonist that is currently being analyzed in a large phase 2 clinical trial for the topical treatment of androgenetic alopecia (AGA). While the pathogenesis of AGA is still debated, the consensus is that AGA is an androgen-dependent hair disorder with strong genetic links, and that the testosterone metabolite, dihydrotestosterone (DHT), plays a causal role in its development. DHT binds to the androgen receptor (AR) in scalp dermal papilla cells (DPC) to induce AR-mediated transcription of genes that contribute to AGA in genetically predisposed individuals. Several studies have established that clascoterone is a potent antiandrogen that is well tolerated and has selective topical activity. The study described herein elucidates a potential mechanism of clascoterone in AGA. Clascoterone was found to inhibit AR-regulated transcription in a reporter cell line with similar efficacy to the 5α-reductase inhibitor, finasteride. More importantly, when compared with another direct AR antagonist, enzalutamide, clascoterone was significantly better at inhibiting IL-6 synthesis from DHT-stimulated primary cultures of human scalp DPC. Therefore, clascoterone may be an excellent candidate to be the first topical antiandrogen for treating AGA. J Drugs Dermatol. 2019;18(2):197-201.
Sigma-Aldrich
Clascoterone, ≥98% (HPLC)