Merck
  • ChrR, a soluble quinone reductase of Pseudomonas putida that defends against H2O2.
ChrR, a soluble quinone reductase of Pseudomonas putida that defends against H2O2.
The Journal of biological chemistry (2005-04-21)
Claudio F Gonzalez, David F Ackerley, Susan V Lynch, A Matin
ABSTRACT
Most bacteria contain soluble quinone-reducing flavoenzymes. However, no biological benefit for this activity has previously been demonstrated. ChrR of Pseudomonas putida is one such enzyme that has also been characterized as a chromate reductase; yet we propose that it is the quinone-reducing activity of ChrR that has the greatest biological significance. ChrR reduces quinones by simultaneous two-electron transfer, avoiding formation of highly reactive semiquinone intermediates and producing quinols that promote tolerance of H(2)O(2). Expression of chrR was induced by H(2)O(2), and levels of chrR expression in overexpressing, wild type, and knock-out mutant strains correlated with the H(2)O(2) tolerance and scavenging ability of each strain. The chrR expression level also correlated with intracellular H(2)O(2) levels as measured by protein carbonylation assays and fluorescence-activated cell scanning analysis with the H(2)O(2)-responsive dye H(2)DCFDA. Thus, enhancing the activity of ChrR in a chromate-remediating bacterial strain may not only increase the rate of chromate transformation, it may also augment the capacity of these cells to withstand the unavoidable production of H(2)O(2) that accompanies chromate reduction.
Sigma-Aldrich
Diaphorase from Clostridium kluyveri, recombinant, expressed in E. coli, Animal-component free
Pricing and availability is not currently available.