Merck
  • Observation and Analysis of RAD51 Nucleation Dynamics at Single-Monomer Resolution.
Observation and Analysis of RAD51 Nucleation Dynamics at Single-Monomer Resolution.
Methods in enzymology (2018-02-21)
Shyamal Subramanyam, Colin D Kinz-Thompson, Ruben L Gonzalez, Maria Spies
ABSTRACT
Human RAD51 promotes accurate DNA repair by homologous recombination and is involved in protection and repair of damaged DNA replication forks. The active species of RAD51 and related recombinases in all organisms is a nucleoprotein filament assembled on single-stranded DNA (ssDNA). The formation of a nucleoprotein filament competent for the recombination reaction, or for DNA replication support, is a delicate and strictly regulated process, which occurs through filament nucleation followed by filament extension. The rates of these two phases of filament formation define the capacity of RAD51 to compete with the ssDNA-binding protein RPA, as well as the lengths of the resulting filament segments. Single-molecule approaches can provide a wealth of quantitative information on the kinetics of RAD51 nucleoprotein filament assembly, internal dynamics, and disassembly. In this chapter, we describe how to set up a single-molecule total internal reflection fluorescence microscopy experiment to monitor the initial steps of RAD51 nucleoprotein filament formation in real-time and at single-monomer resolution. This approach is based on the unique, stretched-ssDNA conformation within the recombinase nucleoprotein filament and follows the efficiency of Förster resonance energy transfer (EFRET) between two DNA-conjugated fluorophores. We will discuss the practical aspects of the experimental setup, extraction of the FRET trajectories, and how to analyze and interpret the data to obtain information on RAD51 nucleation kinetics, the mechanism of nucleation, and the oligomeric species involved in filament formation.
Sigma-Aldrich
(R)-(+)-6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic acid, 98%
Pricing and availability is not currently available.