Merck
  • Harnessing a Transient Gene Expression System in Nicotiana benthamiana to Explore Plant Agrochemical Transporters.
Harnessing a Transient Gene Expression System in Nicotiana benthamiana to Explore Plant Agrochemical Transporters.
Plants (Basel, Switzerland) (2021-04-04)
Bingqi Wu, Zhiting Chen, Xiaohui Xu, Ronghua Chen, Siwei Wang, Hanhong Xu, Fei Lin
ABSTRACT
Functional characterization of plant agrichemical transporters provided an opportunity to discover molecules that have a high mobility in plants and have the potential to increase the amount of pesticides reaching damage sites. Agrobacterium-mediated transient expression in tobacco is simple and fast, and its protein expression efficiency is high; this system is generally used to mediate heterologous gene expression. In this article, transient expression of tobacco nicotine uptake permease (NtNUP1) and rice polyamine uptake transporter 1 (OsPUT1) in Nicotiana benthamiana was performed to investigate whether this system is useful as a platform for studying the interactions between plant transporters and pesticides. The results showed that NtNUP1 increases nicotine uptake in N. benthamiana foliar discs and protoplasts, indicating that this transient gene expression system is feasible for studying gene function. Moreover, yeast expression of OsPUT1 apparently increases methomyl uptake. Overall, this method of constructing a transient gene expression system is useful for improving the efficiency of analyzing the functions of plant heterologous transporter-encoding genes and revealed that this system can be further used to study the functions of transporters and pesticides, especially their interactions.
Product Description
USP
Nicotine Related Compound F, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.