Merck
In situ analysis of gelatinolytic activity in human dentin.
Acta histochemica (2018-01-27)
Thiago Henrique Scarabello Stape, Leo Tjäderhane, Arzu Tezvergil-Mutluay, Wagner Gomes Da Silva, Alan Roger Dos Santos Silva, Wander José da Silva, Marcelo Rocha Marques
ABSTRACT
Matrix metalloproteinases (MMPs) such as gelatinases are differentially expressed in human tissues. These enzymes cleave specific substrates involved in cell signaling, tissue development and remodeling and tissue breakdown. Recent evidences show that gelatinases are crucial for normal dentin development and their activity is maintained throughout the entire tooth function in the oral cavity. Due to the lack of information about the exact location and activity of gelatinases in mature human dentin, the present study was designed to examine gelatinolytic levels in sound dentin. In situ zymography using confocal microscopy was performed on both mineralized and demineralized dentin samples. Sites presenting gelatinase activity were identified throughout the entire biological tissue pursuing different gelatinolytic levels for distinct areas: predentin and dentinal tubule regions presented higher gelatinolytic activity compared to intertubular dentin. Dentin regions with higher gelatinolytic activity immunohistochemically were partially correlated with MMP-2 expression. The maintenance of gelatinolytic activity in mature dentin may have biological implications related to biomineralization of predentin and tubular/peritubular dentinal regions, as well as regulation of defensive mechanisms of the dentin-pulp complex.
Sigma-Aldrich
Anti-MMP-2 (Ab-3) Mouse mAb (42-5D11), liquid, clone 42-5D11, Calbiochem®