Merck
  • NAC blocks Cystatin C amyloid complex aggregation in a cell system and in skin of HCCAA patients.
NAC blocks Cystatin C amyloid complex aggregation in a cell system and in skin of HCCAA patients.
Nature communications (2021-03-25)
Michael E March, Alvaro Gutierrez-Uzquiza, Asbjorg Osk Snorradottir, Leticia S Matsuoka, Noelia Fonseca Balvis, Thorgeir Gestsson, Kenny Nguyen, Patrick M A Sleiman, Charlly Kao, Helgi J Isaksson, Birkir Thor Bragason, Elias Olafsson, Astridur Palsdottir, Hakon Hakonarson
ABSTRACT
Hereditary cystatin C amyloid angiopathy is a dominantly inherited disease caused by a leucine to glutamine variant of human cystatin C (hCC). L68Q-hCC forms amyloid deposits in brain arteries associated with micro-infarcts, leading ultimately to paralysis, dementia and death in young adults. To evaluate the ability of molecules to interfere with aggregation of hCC while informing about cellular toxicity, we generated cells that produce and secrete WT and L68Q-hCC and have detected high-molecular weight complexes formed from the mutant protein. Incubations of either lysate or supernatant containing L68Q-hCC with reducing agents glutathione or N-acetyl-cysteine (NAC) breaks oligomers into monomers. Six L68Q-hCC carriers taking NAC had skin biopsies obtained to determine if hCC deposits were reduced following NAC treatment. Remarkably, ~50-90% reduction of L68Q-hCC staining was observed in five of the treated carriers suggesting that L68Q-hCC is a clinical target for reducing agents.
Sigma-Aldrich
Anti-CST3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.