Merck
  • Stroke subtype-dependent synapse elimination by reactive gliosis in mice.
Stroke subtype-dependent synapse elimination by reactive gliosis in mice.
Nature communications (2021-11-28)
Xiaojing Shi, Longlong Luo, Jixian Wang, Hui Shen, Yongfang Li, Muyassar Mamtilahun, Chang Liu, Rubing Shi, Joon-Hyuk Lee, Hengli Tian, Zhijun Zhang, Yongting Wang, Won-Suk Chung, Yaohui Tang, Guo-Yuan Yang
ABSTRACT
The pathological role of reactive gliosis in CNS repair remains controversial. In this study, using murine ischemic and hemorrhagic stroke models, we demonstrated that microglia/macrophages and astrocytes are differentially involved in engulfing synapses in the reactive gliosis region. By specifically deleting MEGF10 and MERTK phagocytic receptors, we determined that inhibiting phagocytosis of microglia/macrophages or astrocytes in ischemic stroke improved neurobehavioral outcomes and attenuated brain damage. In hemorrhagic stroke, inhibiting phagocytosis of microglia/macrophages but not astrocytes improved neurobehavioral outcomes. Single-cell RNA sequencing revealed that phagocytosis related biological processes and pathways were downregulated in astrocytes of the hemorrhagic brain compared to the ischemic brain. Together, these findings suggest that reactive microgliosis and astrogliosis play individual roles in mediating synapse engulfment in pathologically distinct murine stroke models and preventing this process could rescue synapse loss.
Sigma-Aldrich
Anti-Megf10 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-LAMP-2 Antibody, clone ABL-93, clone ABL-93, from rat
Sigma-Aldrich
Anti-MAP2 Antibody, clone AP20, clone AP20, Chemicon®, from mouse