Merck
  • Extracellular matrix protein N-glycosylation mediates immune self-tolerance in Drosophila melanogaster.
Extracellular matrix protein N-glycosylation mediates immune self-tolerance in Drosophila melanogaster.
Proceedings of the National Academy of Sciences of the United States of America (2021-09-22)
Nathan T Mortimer, Mary L Fischer, Ashley L Waring, Pooja Kr, Balint Z Kacsoh, Susanna E Brantley, Erin S Keebaugh, Joshua Hill, Chris Lark, Julia Martin, Pravleen Bains, Jonathan Lee, Alysia D Vrailas-Mortimer, Todd A Schlenke
ABSTRACT
In order to respond to infection, hosts must distinguish pathogens from their own tissues. This allows for the precise targeting of immune responses against pathogens and also ensures self-tolerance, the ability of the host to protect self tissues from immune damage. One way to maintain self-tolerance is to evolve a self signal and suppress any immune response directed at tissues that carry this signal. Here, we characterize the Drosophila tuSz 1 mutant strain, which mounts an aberrant immune response against its own fat body. We demonstrate that this autoimmunity is the result of two mutations: 1) a mutation in the GCS1 gene that disrupts N-glycosylation of extracellular matrix proteins covering the fat body, and 2) a mutation in the Drosophila Janus Kinase ortholog that causes precocious activation of hemocytes. Our data indicate that N-glycans attached to extracellular matrix proteins serve as a self signal and that activated hemocytes attack tissues lacking this signal. The simplicity of this invertebrate self-recognition system and the ubiquity of its constituent parts suggests it may have functional homologs across animals.
Sigma-Aldrich
Anti-MOGS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution