- The role of HIV Tat protein in HIV-related cardiovascular diseases.
The role of HIV Tat protein in HIV-related cardiovascular diseases.
Journal of translational medicine (2018-05-10)
Yanan Jiang, Lu Chai, Moyondafoluwa Blessing Fasae, Yunlong Bai
PMID29739413
ABSTRACT
The human immunodeficiency virus (HIV) is a major global public health issue. HIV-related cardiovascular disease remains a leading cause of morbidity and mortality in HIV positive patients. HIV Tat is a regulatory protein encoded by tat gene of HIV-1, which not only promotes the transcription of HIV, but it is also involved in the pathogenesis of HIV-related complications. This review is aimed at summarizing the current understanding of Tat in HIV-related cardiovascular diseases.