Merck
Plasma mevalonate concentrations in uremic patients.
Kidney international (1997-03-01)
A Scoppola, P De Paolis, G Menzinger, A Lala, S Di Giulio
ABSTRACT
Mevalonic acid (mevalonate or MVA), is an obligate precursor in the biosynthetic pathway of cholesterol. It is partially metabolized by the kidneys and its plasma concentrations are an index of endogenous cholesterol synthesis. The aim of the present study was to evaluate plasma MVA concentrations in uremic patients with different degrees of chronic renal failure (CRF; group A), and the effects of a single hemodialysis treatment on plasma MVA in a group of patients with end-stage renal disease (ESRD; group B). CRF patients exhibited a higher mean basal mevalonate concentration (13.3 +/- 6.5 ng/ml) than control subjects (4.68 +/- 1.32 ng/ml; P < 0.001). A statistically significant direct correlation was evident in CRF patients between mevalonate and creatinine plasma levels (r = 0.86; P < 0.001). A single hemodialysis treatment was associated with a significant reduction of plasma mevalonate concentrations four hours after the hemodialysis session (-57%; P < 0.001) and an increase up to the basal values 24 hours after the end of the treatment. In conclusion, our results demonstrated: (i) higher plasma MVA concentrations in patients with decreased renal function; (ii) a direct relationship between plasma MVA levels and the degree of kidney failure as expressed by creatinine plasma concentrations; and (iii) a clear cut reduction of elevated plasma MVA levels after a single hemodialysis treatment.
Sigma-Aldrich
(R)-Mevalonic acid lithium salt, ≥93.0% (qNMR)